Glutarimid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Piperidine-2,6-dione
L00968.06
L00968.14
L00968.06EA
42.1
EUR
L00968.06
L00968.14
Glutarimid ≥98%
Glutarimid
Purity: 98%
CAS No.: 1121-89-7
Molecular Formula: C5H7NO2
Formula Weight: 113.12
Odour: Nearly Odourless
Melting Point: 153-157°
Beilstein Reference: 110052
MDL No.: MFCD00006670
Formel:
C₅H₇NO₂ Molecular Weight: 113.12 g/mol Schmelzpunkt: 153…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006670 CAS-Nummer: 1121-89-7 EINECS: 214-340-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...