Octadecylsilan

Lieferant: CHEMPUR SILANE

SILS12240.10 SILS12240.25 SILS12240.100
SILNSILS12240.10EA 0 EUR
SILNSILS12240.10 SILNSILS12240.25 SILNSILS12240.100
Octadecylsilan
Octadecylsilane
Formel: C₁₈H₄₀Si
Molecular Weight: 284.60 g/mol
MDL: MFCD00039874
CAS-Nummer: 18623-11-5
EINECS: 242-453-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR