Octylsilan
Lieferant: CHEMPUR SILANE
SILS12725.10
SILS12725.25
SILNSILS12725.10EA
0
EUR
SILNSILS12725.10
SILNSILS12725.25
Octylsilan
Octylsilane
Formel:
C₈H₂₀Si |
CAS-Nummer:
871-92-1 EINECS: 212-817-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...