Didecyldimethylammoniumbromid ≥80%, wässriges Gel
Lieferant: Thermo Scientific
|
Achtung
|
B24832.22
B24832.36
B24832.22EA
46.6
EUR
B24832.22
B24832.36
Didecyldimethylammoniumbromid ≥80%, wässriges Gel
Didecyldimethylammonium bromide
CAS No.: 2390-68-3
Molecular Formula: C22H48BrN
Formula Weight: 406.54
Refractive Index: 1.4570
MDL No.: MFCD00012194
Formel:
C₂₂H₄₈BrN Molecular Weight: 406,54 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012194 CAS-Nummer: 2390-68-3 EINECS: 219-234-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...