Didodecyldimethylammoniumbromid 99%
Lieferant: Thermo Scientific
|
Achtung
|
407121000
407120250
ACRO407121000EA
415
EUR
ACRO407121000
ACRO407120250
Didodecyldimethylammoniumbromid 99%
Didodecyldimethylammonium bromide
Formel:
C₂₆H₅₆BrN Molecular Weight: 462,65 g/mol Schmelzpunkt: 161…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041969 CAS-Nummer: 3282-73-3 EINECS: 221-923-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 158 to 163 °C |
Titration Argentometric | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...