2,4,6-Tribrom-3-nitrophenol

Lieferant: COMBI-BLOCKS

AN-1425-5G AN-1425-1G
COBBAN-1425-5GEA 149 EUR
COBBAN-1425-5G COBBAN-1425-1G
2,4,6-Tribrom-3-nitrophenol
2,4,6-Tribrom-3-nitrophenol
Formel: C₆H₂Br₃NO₃
MDL: MFCD12025953
CAS-Nummer: 69076-62-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR