Trihexyl(tetradecyl)phosphoniumchlorid, Sigma-Aldrich®

Lieferant: Merck

89744-5G-F 89744-50G-F
SIAL89744-5G-FEA 48.2 EUR
SIAL89744-5G-F SIAL89744-50G-F
Trihexyl(tetradecyl)phosphoniumchlorid, Sigma-Aldrich®
Trihexyl(tetradecyl)phosphoniumchlorid
Formel: C₃₂H₆₈ClP
Molecular Weight: 519,31 g/mol
MDL: MFCD04112746
CAS-Nummer: 258864-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR