1-Chlor-3-(trifluormethoxy)benzol
Lieferant: Thermo Scientific
Synonyme:
3-(Trifluoromethoxy)chlorobenzene
TL00820.1
TL00820.10
MAYBTL00820.1EA
0
EUR
MAYBTL00820.1
MAYBTL00820.10
1-Chlor-3-(trifluormethoxy)benzol
1-Chloro-3-(trifluoromethoxy)benzene
Formel:
C₇H₄ClF₃O Molecular Weight: 196.55 g/mol |
MDL:
MFCD00276971 CAS-Nummer: 772-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...