(±)-2-Methyltetrahydrofuran-3-on

Lieferant: Thermo Scientific

Synonyme: 2-Methyltetrahydrofuran-3-one

SB01113DA SB01113EA SB01113.50
MAYBSB01113DAEA 0 EUR
MAYBSB01113DA MAYBSB01113EA MAYBSB01113.50
(±)-2-Methyltetrahydrofuran-3-on
(±)-2-Methyltetrahydrofuran-3-on
Formel: C₅H₈O₂
Molecular Weight: 100,12 g/mol
MDL: MFCD00010423
CAS-Nummer: 3188-00-9
EINECS: 221-685-4
UN: 1224
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR