(±)-2-Methyltetrahydrofuran-3-on, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Methyltetrahydrofuran-3-one
M-Clarity™ Qualitätsstufe = MQ300
W337307-100G-K
W337307-1KG-K
W337307-250G-K
W337307-5KG-K
SIALW337307-100G-KEA
186
EUR
SIALW337307-100G-K
SIALW337307-1KG-K
SIALW337307-250G-K
SIALW337307-5KG-K
(±)-2-Methyltetrahydrofuran-3-on, Sigma-Aldrich®
(±)-2-Methyltetrahydrofuran-3-on
Formel:
C₅H₈O₂ Molecular Weight: 100,12 g/mol |
MDL:
MFCD00010423 CAS-Nummer: 3188-00-9 EINECS: 221-685-4 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...