Zirkonium(IV)ethanolat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

339121-25G 339121-5G
SIAL339121-25GEA 286 EUR
SIAL339121-25G SIAL339121-5G
Zirkonium(IV)ethanolat, Sigma-Aldrich®
Zirkonium(IV)ethanolat
Formel: C₈H₂₀O₄Zr
Molecular Weight: 271,47 g/mol
MDL: MFCD00015654
CAS-Nummer: 18267-08-8
UN: 3181
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR