(Hydroxypropyl)methylcellulose (Hypromellose)
Lieferant: Thermo Fisher Scientific
Synonyme: Methocel K100, Hydroxypropyl methyl cellulose
44779.18 44779.30 44779.A1
44779.18EA 45.8 EUR
44779.18 44779.30 44779.A1
(Hydroxypropyl)methylcellulose (Hypromellose)
(Hydroxypropyl)methylcellulose (Hypromellose)
CAS No.: 9004-65-3 Physical Form: Powder Merck Reference: 14,4842 MDL No.: MFCD00131360
Bitte beachten Incompatible with strong oxidising agents, bleaching powder, fluorine and nitric acid.
Formel: C₃H₈O₂·xCH₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00131360
CAS-Nummer: 9004-65-3
Merck Index: 14,4842