2,2,2-Trifluor-N-phenylacetamid

Lieferant: BIONET RESEARCH

8E-016.500 8E-016.1
BIOT8E-016.500EA 0 EUR
BIOT8E-016.500 BIOT8E-016.1
2,2,2-Trifluor-N-phenylacetamid
2,2,2-Trifluor-N-phenylacetamid
Formel: C₈H₆F₃NO
Molecular Weight: 189.1346 g/mol
MDL: MFCD00196027
CAS-Nummer: 404-24-0
EINECS: 206-965-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR