2,2,2-Trifluor-N-phenylacetamid

Lieferant: FLUOROCHEM

010745-1G 010745-5G
FLUO010745-1GEA 51.6 EUR
FLUO010745-1G FLUO010745-5G
2,2,2-Trifluor-N-phenylacetamid
2,2,2-Trifluor-N-phenylacetamid
Formel: C₈H₆F₃NO
Molecular Weight: 189,1346 g/mol
MDL: MFCD00196027
CAS-Nummer: 404-24-0
EINECS: 206-965-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR