3-Brom-5-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

661112-500MG 661112-5G
SIAL661112-500MGEA 85.9 EUR
SIAL661112-500MG SIAL661112-5G
3-Brom-5-(trifluormethyl)pyridin, Sigma-Aldrich®
3-Bromo-5-(trifluoromethyl)pyridine
Formel: C₆H₃BrF₃N
Molecular Weight: 226,00 g/mol
MDL: MFCD04972700
CAS-Nummer: 436799-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR