5-Brom-2-(trifluormethyl)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-trifluoromethylpyridine

OS-7412-5G OS-7412-25G OS-7412-1G
COBBOS-7412-5GEA 66 EUR
COBBOS-7412-5G COBBOS-7412-25G COBBOS-7412-1G
5-Brom-2-(trifluormethyl)pyridin
5-Brom-2-(trifluormethyl)pyridin
Formel: C₆H₃BrF₃N
Molecular Weight: 226,00 g/mol
MDL: MFCD06657686
CAS-Nummer: 436799-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR