5-Brom-2-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

Synonyme: 5-Bromo-2-trifluoromethylpyridine

013952-1G 013952-5G
FLUO013952-1GEA 0 EUR
FLUO013952-1G FLUO013952-5G
5-Brom-2-(trifluormethyl)pyridin
5-Brom-2-(trifluormethyl)pyridin
Formel: C₆H₃BrF₃N
Molecular Weight: 226.00 g/mol
MDL: MFCD06657686
CAS-Nummer: 436799-32-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR