5-Brom-2-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromo-2-trifluoromethylpyridine

M-Clarity™ Qualitätsstufe = MQ200

661104-500MG 661104-5G
SIAL661104-500MGEA 76.6 EUR
SIAL661104-500MG SIAL661104-5G
5-Brom-2-(trifluormethyl)pyridin, Sigma-Aldrich®
5-Brom-2-(trifluormethyl)pyridin
Formel: C₆H₃BrF₃N
Molecular Weight: 226,00 g/mol
MDL: MFCD06657686
CAS-Nummer: 436799-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR