(Pentafluorethyl)trimethylsilan
Lieferant: CHEMPUR SILANE
SILS12830.1
SILS12830.5
SILNSILS12830.1EA
0
EUR
SILNSILS12830.1
SILNSILS12830.5
(Pentafluorethyl)trimethylsilan
(Pentafluoroethyl)trimethylsilane
Formel:
C₅H₉F₅Si Molecular Weight: 192.21 g/mol |
MDL:
MFCD04116436 CAS-Nummer: 124898-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...