2-Fluor-4-hydroxybenzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

H26218.MC H26218.ME
H26218.MCEA 93.4 EUR
H26218.MC H26218.ME
2-Fluor-4-hydroxybenzaldehyd ≥97%
2-Fluoro-4-hydroxybenzaldehyde

Purity: 97% CAS No.: 348-27-6 Molecular Formula: C7H5FO2 Formula Weight: 140.11 Melting Point: 168-170° MDL No.: MFCD06797918

Formel: C₇H₅FO₂
Molecular Weight: 140,11 g/mol
Schmelzpunkt: 168…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797918
CAS-Nummer: 348-27-6
EINECS: None

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR