N-Acetyl-α-D-glucosamin 99+%

Lieferant: Thermo Scientific

Synonyme: 2-Acetamido-2-deoxy-α-D-glucopyranose

102560250 102561000
ACRO102560250EA 62 EUR
ACRO102560250 ACRO102561000
N-Acetyl-α-D-glucosamin 99+%
N-Acetyl-α-D-glucosamin
Formel: C₈H₁₅NO₆
Schmelzpunkt: 211 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00064359
CAS-Nummer: 10036-64-3
EINECS: 231-368-2

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1.0 % (105°C, 4 h)
Sulfated ash ≤0.5 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation +39° to +43° (20°C, 589 nm) (c=2, H2O, after 2 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR