N-Acetyl-β-D-glucosamin
Lieferant: Cayman Chemical
Synonyme:
2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine
13136-500
13136-1
13136-5
13136-10
CAYM13136-500EA
19
EUR
CAYM13136-500
CAYM13136-1
CAYM13136-5
CAYM13136-10
N-Acetyl-β-D-glucosamin
N-Acetyl-β-D-glucosamin
Formel:
C₈H₁₅NO₆ Molecular Weight: 221,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00136044 CAS-Nummer: 7512-17-6 EINECS: 231-368-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...