N-Acetyl-β-D-glucosamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine

M-Clarity™ Qualitätsstufe = MQ200

A4106-100G A4106-25G A4106-500G
SIALA4106-100GEA 209 EUR
SIALA4106-100G SIALA4106-25G SIALA4106-500G
N-Acetyl-β-D-glucosamin, Sigma-Aldrich®
N-Acetyl-β-D-glucosamin
Formel: C₈H₁₅NO₆
Molecular Weight: 221,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00136044
CAS-Nummer: 7512-17-6
EINECS: 231-368-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR