2-Desoxy-2-fluor-D-glucose, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Fluoro-2-deoxy-D-glucose, 2-Deoxy-2-fluoro-D-glucopyranose, FDG
M-Clarity™ Qualitätsstufe = MQ300
F5006-100MG
F5006-25MG
SIALF5006-100MGEA
1020
EUR
SIALF5006-100MG
SIALF5006-25MG
2-Desoxy-2-fluor-D-glucose, Sigma-Aldrich®
2-Deoxy-2-fluoro-D-glucose
Formel:
C₆H₁₁FO₅ Molecular Weight: 182,15 g/mol |
MDL:
MFCD00077527 CAS-Nummer: 29702-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...