2-Desoxy-D-glucose ≥98%

Lieferant: Thermo Scientific

Synonyme: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal

L07338.03 L07338.06
L07338.03EA 33.2 EUR
L07338.03 L07338.06
2-Desoxy-D-glucose ≥98%
2-Deoxy-D-glucose

Purity: 98% CAS No.: 154-17-6 Molecular Formula: C6H12O5 Formula Weight: 164.16 Melting Point: 147-152° Beilstein Reference: 1723331 Merck Reference: 14,2904 Optical Rotation: +47° (c=0.5 in water, 45h) MDL No.: MFCD00151328

Formel: C₆H₁₂O₅
Molecular Weight: 164,16 g/mol
Schmelzpunkt: 147…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151328
CAS-Nummer: 154-17-6
EINECS: 205-823-0
Merck Index: 14,2904

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR