2-Desoxy-D-glucose, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal

M-Clarity™ Qualitätsstufe = MQ200

D8375-100G D8375-10MG D8375-1G D8375-25G D8375-5G
SIALD8375-100GEA 3110 EUR
SIALD8375-100G SIALD8375-10MG SIALD8375-1G SIALD8375-25G SIALD8375-5G
2-Desoxy-D-glucose, Sigma-Aldrich®
2-Deoxy-D-glucose
Formel: C₆H₁₂O₅
Molecular Weight: 164,16 g/mol
MDL: MFCD00151328
CAS-Nummer: 154-17-6
EINECS: 205-823-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR