2,4,6-Tribrom-3-(trifluormethyl)phenol

Lieferant: COMBI-BLOCKS

OT-1009-5G OT-1009-1G
COBBOT-1009-5GEA 434 EUR
COBBOT-1009-5G COBBOT-1009-1G
2,4,6-Tribrom-3-(trifluormethyl)phenol
2,4,6-Tribrom-3-(trifluormethyl)phenol
Formel: C₇H₂Br₃F₃O
MDL: MFCD13195740
CAS-Nummer: 384-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR