m-Kresolpurpur
Lieferant: Thermo Scientific
Synonyme:
2',2''-Dimethylphenolsulphonphthalein, m-Cresolsulphonphthalein, MCP, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
B24338.06
B24338.14
B24338.06EA
44.2
EUR
B24338.06
B24338.14
m-Kresolpurpur
m-Kresolpurpur
CAS No.: 2303-01-7
Molecular Formula: C21H18O5S
Formula Weight: 382.44
Beilstein Reference: 350314
Merck Reference: 14,2577
MDL No.: MFCD00005871
Formel:
C₂₁H₁₈O₅S Molecular Weight: 382,43 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005871 CAS-Nummer: 2303-01-7 EINECS: 218-960-6 Merck Index: 14,2577 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...