β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂) Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme: beta-NADH disodium salt hydrate, beta-Nicotinamide adenine dinucleotide disodium salt hydrate, β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form) hydrate
N6005-25G N6005-5G
SIALN6005-25GEA 2230 EUR
SIALN6005-25G SIALN6005-5G
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂) Hydrat, Sigma-Aldrich®
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na2) Hydrat
Formel: C₂₁H₂₇N₇Na₂O₁₄P₂·H₂O
Molecular Weight: 727,43 g/mol
MDL: MFCD03791273
CAS-Nummer: 606-68-8
EINECS: 210-123-3