1,2-Dichlor-3-fluorbenzol
Lieferant: FLUOROCHEM
Synonyme:
2,3-Dichlorofluorobenzene
004706-1G
004706-5G
FLUO004706-1GEA
31.1
EUR
FLUO004706-1G
FLUO004706-5G
1,2-Dichlor-3-fluorbenzol
1,2-Dichloro-3-fluorobenzene
Formel:
C₆H₃Cl₂F Molecular Weight: 164,9924 g/mol |
MDL:
MFCD00060647 CAS-Nummer: 36556-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...