1,3-Dichlor-5-fluorbenzol
Lieferant: FLUOROCHEM
Synonyme:
3,5-Dichlorofluorobenzene
004709-5G
004709-10G
FLUO004709-5GEA
45.8
EUR
FLUO004709-5G
FLUO004709-10G
1,3-Dichlor-5-fluorbenzol
1,3-Dichloro-5-fluorobenzene
Formel:
C₆H₃Cl₂F Molecular Weight: 164,9900 g/mol |
MDL:
MFCD00060652 CAS-Nummer: 1435-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...