1-Chlor-2,6-difluorbenzol

Lieferant: CHEMPUR

FL13251.POR 013251-25G 013251-5G 013251-1G
CHMPFL13251.POREA 0 EUR
CHMPFL13251.POR FLUO013251-25G FLUO013251-5G FLUO013251-1G
1-Chlor-2,6-difluorbenzol
1-Chloro-2,6-difluorobenzene
Formel: C₆H₃ClF₂
MDL: MFCD00142604
CAS-Nummer: 38361-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR