3,5-Di-tert-butyl-2-hydroxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Di-tert-butylsalicylaldehyde

M-Clarity™ Qualitätsstufe = MQ100

140414-25G 140414-5G
SIAL140414-25GEA 53.5 EUR
SIAL140414-25G SIAL140414-5G
3,5-Di-tert-butyl-2-hydroxybenzaldehyd, Sigma-Aldrich®
3,5-Di-tert-butyl-2-hydroxybenzaldehyd
Formel: C₁₅H₂₂O₂
Molecular Weight: 234.34 g/mol
MDL: MFCD00191998
CAS-Nummer: 37942-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR