Spermin Tetrahydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine) tetrahydrochloride
M-Clarity™ Qualitätsstufe = MQ300
85610-25G
85610-5G
SIAL85610-25GEA
631
EUR
SIAL85610-25G
SIAL85610-5G
Spermin Tetrahydrochlorid, Sigma-Aldrich®
Spermin Tetrahydrochlorid
Formel:
C₁₀H₃₀Cl₄N₄ Molecular Weight: 348.19 g/mol |
MDL:
MFCD00012914 CAS-Nummer: 306-67-2 EINECS: 206-189-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...