Spermin Tetrahydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine) tetrahydrochloride

M-Clarity™ Qualitätsstufe = MQ200

S2876-5G S2876-100G S2876-10G S2876-1G S2876-25G
SIALS2876-5GEA 298 EUR
SIALS2876-5G SIALS2876-100G SIALS2876-10G SIALS2876-1G SIALS2876-25G
Spermin Tetrahydrochlorid, Sigma-Aldrich®
Spermin Tetrahydrochlorid
Formel: C₁₀H₃₀Cl₄N₄
Molecular Weight: 348.19 g/mol
MDL: MFCD00012914
CAS-Nummer: 306-67-2
EINECS: 206-189-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR