(3,3,3-Trifluorpropyl)dichlormethylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Dichloro(methyl)(3,3,3-trifluoropropyl)silane, (3,3,3-Trifluoropropyl)methyldichlorosilane

SILS17750.5 SILS17750.25
SILNSILS17750.5EA 0 EUR
SILNSILS17750.5 SILNSILS17750.25
(3,3,3-Trifluorpropyl)dichlormethylsilan
(3,3,3-Trifluorpropyl)dichlormethylsilan
Formel: C₄H₇Cl₂F₃Si
Molecular Weight: 211,09 g/mol
MDL: MFCD00039294
CAS-Nummer: 675-62-7
EINECS: 211-623-4
UN: 2985
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR