(3,3,3-Trifluorpropyl)dimethoxymethylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane, 1,1,1-Trifluoro-3-[dimethoxy(methyl)silyl]propane

SILS17775.5 SILS17775.25
SILNSILS17775.5EA 0 EUR
SILNSILS17775.5 SILNSILS17775.25
(3,3,3-Trifluorpropyl)dimethoxymethylsilan
(3,3,3-Trifluorpropyl)dimethoxymethylsilan
Formel: C₆H₁₃F₃O₂Si
Molecular Weight: 202.25 g/mol
MDL: MFCD00053175
CAS-Nummer: 358-67-8
EINECS: 206-619-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR