1,2-Diethoxy-1,1,2,2-tetramethyldisilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

667897-1G 667897-5G
SIAL667897-1GEA 220 EUR
SIAL667897-1G SIAL667897-5G
1,2-Diethoxy-1,1,2,2-tetramethyldisilan, Sigma-Aldrich®
1,2-Diethoxy-1,1,2,2-tetramethyldisilan
Formel: C₈H₂₂O₂Si₂
CAS-Nummer: 18419-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR