(+)-Di-p-toluoyl-D-weinsäure ≥98%

Lieferant: Thermo Scientific

Synonyme: Di-p-toluoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid, Di-p-toluoyl-D-tartaric Acid

L13223.14 L13223.22
L13223.14EA 77.8 EUR
L13223.14 L13223.22
(+)-Di-p-toluoyl-D-weinsäure ≥98%
(+)-Di-p-toluoyl-D-weinsäure

Purity: 98% CAS No.: 32634-68-7 Molecular Formula: C20H18O8 Formula Weight: 386.36 Melting Point: 169-171° Beilstein Reference: 3225584

Formel: C₂₀H₁₈O₈
Molecular Weight: 386,36 g/mol
Schmelzpunkt: 169…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008552
CAS-Nummer: 32634-68-7
EINECS: 251-132-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR