(+)-Di-p-toluoyl-D-weinsäure

Lieferant: FLUOROCHEM

Synonyme: Di-p-toluoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid, Di-p-toluoyl-D-tartaric Acid

011666-100G 011666-25G
FLUO011666-100GEA 90.9 EUR
FLUO011666-100G FLUO011666-25G
(+)-Di-p-toluoyl-D-weinsäure
(+)-Di-p-toluoyl-D-weinsäure
Formel: C₂₀H₁₈O₈
Molecular Weight: 386,36 g/mol
MDL: MFCD00008552
CAS-Nummer: 32634-68-7
EINECS: 251-132-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR