TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid) ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
TCEP HCl
40587.02
40587.04
40587.09
40587.02EA
49.6
EUR
40587.02
40587.04
40587.09
TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid) ≥98%
TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)
Purity: 98%
CAS No.: 51805-45-9
Molecular Formula: C9H15O6P.HCl
Formula Weight: 286.65
Physical Form: Crystalline powder
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 177-178°
Solubility: Soluble in water
Merck Reference: 14,9085
UN No.: 3261
MDL No.: MFCD00145469
Reducing agent, specific for disulfides
Formel:
C₉H₁₆ClO₆P Schmelzpunkt: 177…178 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145469 CAS-Nummer: 51805-45-9 UN: 3261 ADR: 8,II Merck Index: 14,9085 |