6-Chlor-2-fluorpurin
Lieferant: FLUOROCHEM
Synonyme:
6-Chloro-2-fluoro-9H-purine
009088-100MG
009088-500MG
009088-1G
009088-5G
FLUO009088-100MGEA
0
EUR
FLUO009088-100MG
FLUO009088-500MG
FLUO009088-1G
FLUO009088-5G
6-Chlor-2-fluorpurin
6-Chloro-2-fluoropurine
Formel:
C₅H₂ClFN₄ Molecular Weight: 172,55 g/mol |
MDL:
MFCD02183557 CAS-Nummer: 1651-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...