6-(Dimethylamino)purin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N,N-Dimethyl-7H-purin-6-amine, N6,N6-Dimethyladenine

D2629-100MG D2629-250MG D2629-500MG D2629-1G
SIALD2629-100MGEA 45 EUR
SIALD2629-100MG SIALD2629-250MG SIALD2629-500MG SIALD2629-1G
6-(Dimethylamino)purin, Sigma-Aldrich®
6-(Dimethylamino)purine
Formel: C₇H₉N₅
Molecular Weight: 163,1799 g/mol
MDL: MFCD00005573
CAS-Nummer: 938-55-6
EINECS: 213-344-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR