5-Brompyrimidin-2-carbonitril

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-cyanopyrimidine, 5-bromo-pyrimidine-2-carbonitile

PY-1333-5G PY-1333-1G
COBBPY-1333-5GEA 76.1 EUR
COBBPY-1333-5G COBBPY-1333-1G
5-Brompyrimidin-2-carbonitril
5-Brompyrimidin-2-carbonitril
Formel: C₅H₂BrN₃
Molecular Weight: 183,99 g/mol
MDL: MFCD02940446
CAS-Nummer: 38275-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR