5-Brom-2-(dimethylamino)pyrimidin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-N,N-dimethylpyrimidin-2-amine

OT-8093-5G OT-8093-25G OT-8093-1G
COBBOT-8093-5GEA 111 EUR
COBBOT-8093-5G COBBOT-8093-25G COBBOT-8093-1G
5-Brom-2-(dimethylamino)pyrimidin
5-Bromo-2-(dimethylamino)pyrimidine
Formel: C₆H₈BrN₃
Molecular Weight: 202,0500 g/mol
MDL: MFCD07368228
CAS-Nummer: 38696-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR