5-Brom-2-(methylamino)pyrimidin
Lieferant: COMBI-BLOCKS
Synonyme:
5-Bromo-N-methylpyrimidin-2-amine
OT-8102-5G
OT-8102-25G
OT-8102-1G
COBBOT-8102-5GEA
76.3
EUR
COBBOT-8102-5G
COBBOT-8102-25G
COBBOT-8102-1G
5-Brom-2-(methylamino)pyrimidin
5-Bromo-2-(methylamino)pyrimidine
Formel:
C₅H₆BrN₃ Molecular Weight: 188.0300 g/mol |
MDL:
MFCD07368227 CAS-Nummer: 31402-54-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...