5-Brom-2-(ethylthio)pyrimidin

Lieferant: COMBI-BLOCKS

PY-1278-5G PY-1278-1G
COBBPY-1278-5GEA 149 EUR
COBBPY-1278-5G COBBPY-1278-1G
5-Brom-2-(ethylthio)pyrimidin
5-Bromo-2-(ethylthio)pyrimidine
Formel: C₆H₇BrN₂S
MDL: MFCD13188492
CAS-Nummer: 859941-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR