5-Brom-2-(methylthio)pyrimidin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-methylthiopyrimidine

PY-1108-5G PY-1108-25G PY-1108-1G
COBBPY-1108-5GEA 98.9 EUR
COBBPY-1108-5G COBBPY-1108-25G COBBPY-1108-1G
5-Brom-2-(methylthio)pyrimidin
5-Bromo-2-(methylthio)pyrimidine
Formel: C₅H₅BrN₂S
Molecular Weight: 205,0756 g/mol
MDL: MFCD00187875
CAS-Nummer: 14001-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR