5-Brom-2-(propylthio)pyrimidin

Lieferant: COMBI-BLOCKS

PY-1277-1G PY-1277-5G
COBBPY-1277-1GEA 98.3 EUR
COBBPY-1277-1G COBBPY-1277-5G
5-Brom-2-(propylthio)pyrimidin
5-Bromo-2-(propylthio)pyrimidine
Formel: C₇H₉BrN₂S
MDL: MFCD13195791
CAS-Nummer: 1199773-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR