4,6-Diamino-2-pyrimidinthiol

Lieferant: CHEMPUR

Synonyme: 4,6-Diamino-2-mercaptopyrimidine, 4,6-Diamino-2-pyrimidinethiol

RJF00997.250 RJF00997.1
CHMPRJF00997.250EA 0 EUR
CHMPRJF00997.250 MAYBRJF00997.1
4,6-Diamino-2-pyrimidinthiol
4,6-Diamino-2-pyrimidinthiol
Formel: C₄H₆N₄S
Molecular Weight: 142,18 g/mol
MDL: MFCD00006078
CAS-Nummer: 1004-39-3
EINECS: 213-721-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR